Would you like to log in again?
Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
2110-0006 2110-0008 2110-0016 2110-0002 2110-0032
16120-956CS 437.76 USD InStock
16120-956 16120-953 16120-962 16120-950 16120-959
Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Bottles Roller Bottles
Square, polypropylene wide mouth bottles are designed to replace glass serum and general-purpose storage bottles.
  • Resistant to acids, alcohols, and bases
  • Contact-clear bottles provide visible liquid level
  • Wide mouth for easy cleaning, sample filling, and retrieval
  • Square shape saves space and offers large labeling surface
  • Leakproof and repeatedly autoclavable
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: Supplied with polypropylene screw caps assembled on bottles.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty