Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® Square Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nalgene®
2114-0006 2114-0002 2114-0016 2114-0032 2114-0008
16121-081CS 313.92 USD InStock
16121-081 16120-974 16121-004 16121-048 16121-060
Nalgene® Square Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Roller Bottles
Ideal for liquid or chemical storage, these HDPE square bottles make excellent sample mailers.

  • Square shape saves space
  • Bottles can withstand temperatures up to 120 °C (248 °F)
  • Leakproof, Not autoclavable

Wide mouth design allows for easy filling and dispensing.

Ordering information: Supplied with polypropylene screw caps assembled on bottles.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty