Would you like to log in again?
Nalgene® Square Laboratory Bottles, Polycarbonate, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2015-0250 2015-0500 2015-2000 2015-1000 2015-0060 2015-0125 2015-0030
16121-157CS 545.28 USD InStock
16121-157 16121-159 16121-230 16121-220 16121-208 16121-183 16121-161
Nalgene® Square Laboratory Bottles, Polycarbonate, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Ideal for general-purpose storage of aqueous solutions and short-term media storage.
  • 2 L size has handgrips for easier handling
  • Leakproof
  • Autoclavable
Note: Before autoclaving, set cap on top of the bottle without engaging the threads.
Square design saves space on benchtop or shelf, in refrigerators and freezers, and under the lab hood. Wide square base provides stability. Permanent graduations in convenient increments for easy measuring. Deep cap and narrow mouth design facilitate aseptic technique.
Ordering information: Supplied with polypropylene screw caps assembled on bottles. For replacement caps for 16121-161 through -230, see 16219-122 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty