Would you like to log in again?
Nalgene® Wide-Mouth EP Tox Bottle, Teflon® Resin FEP, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2101-2200
16124-970CS 1056 USD InStock
16124-970
Nalgene® Wide-Mouth EP Tox Bottle, Teflon® Resin FEP, Thermo Scientific
Bottles Water Sampling Bottles
Specially designed for EPA Method 1311: Toxicity Characteristic Leaching Procedure (TCLP).
Note: Before autoclaving, set closure on top of the container without engaging the threads.
Constructed of Teflon® FEP for excellent chemical and temperature resistance. With leakproof Teflon® resin-lined polypropylene cap. Clear, autoclavable bottle can be used at high or low temperatures for trace metal analysis and applications with organic solvents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty