Would you like to log in again?
Nalgene® Laboratory Bottles, Low-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2103-0032 2103-0004 2103-0016 2103-0001 2103-0002 2103-0008
16124-988CS 336.43 USD InStock
16124-988 16124-986 16124-984 16124-982 16124-980 16124-990
Nalgene® Laboratory Bottles, Low-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
View liquid levels clearly in the impact resistant Thermo Scientific™ Nalgene™ Wide-Mouth LDPE bottles with closure.
  • Ideal for storing chemicals and standards, as well as collecting and storing distilled water
  • Extremely low trace metal content in LDPE makes it ideal for trace metal analysis
  • Excellent chemical resistance to most acids, bases and alcohols
  • Wide mouth is easy to fill
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty