Would you like to log in again?
Nalgene® Large Sample Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2120-0010 2120-0005
16125-129CS 212.16 USD InStock
16125-129 16125-118
Nalgene® Large Sample Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Widely used for collecting specimens and water sampling.
  • Leakproof
  • With polypropylene screw closure
Also excellent for use with dry chemicals. Mouth opening: 89 mm (3¹/₂").
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty