Would you like to log in again?
Nalgene® Laboratory Bottles, Polymethylpentene, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2107-0008 2107-0016
16125-424CS 602.88 USD InStock
16125-424 16125-446
Nalgene® Laboratory Bottles, Polymethylpentene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Completely transparent, chemical-resistant plastic bottle is ideal for tissue specimens that must be shipped wet.
  • Withstands temperatures from 20 to 135 °C (68 to 275 °F)
  • With polypropylene closure
  • Leakproof
  • Autoclavable
For replacement caps, see 16219-030 series.
Note: Before autoclaving, set closure on top of bottle without engaging threads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty