Would you like to log in again?
Corning® Polypropylene Snap-Seal Sample Containers, Corning
Supplier: Corning
Corning®
1730-8 1730-4H 1730-10 1730-4L 1730-5X 1730-2C
16126-018CS 161.24 USD InStock
16126-018 16126-010 16126-022 16126-030 16126-014 16126-026
Corning® Polypropylene Snap-Seal Sample Containers, Corning
Sample Containers General Purpose Containers
PP, disposable, translucent.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both millilitres and ounces
  • Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty