Would you like to log in again?
SP Bel-Art Wide Mouth Storage Bottles, Heavy-Duty Closure, LDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F10906-0050 F10906-2000 F10906-0200 F10906-1500 F10906-1000 F10906-0250 F10906-0100 109060010 F10906-0500 F10906-5000
47750-656EA 126.4 USD InStock
47750-656 16126-358 16126-314 16126-176 16126-198 16126-212 16126-234 16126-278 70680-364 70680-366
SP Bel-Art Wide Mouth Storage Bottles, Heavy-Duty Closure, LDPE, Bel-Art Products, a part of SP
Bottles Roller Bottles
Low-density polyethylene bottles with a heavy-threaded, self-sealing screw cap closure provide reliability when storing, shipping, and handling materials in both liquid and powder form.
  • LDPE construction
  • Heavy-threaded, self-sealing screw cap closure
  • Reliable for use with both liquid and powder contents
  • Larger sizes are made of HDPE
Note: 5000 and 10000 ml sizes are made of high-density polyethylene and have an octagonal shape. They are not leakproof and are best used for dry materials.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty