Would you like to log in again?
SP Bel-Art Round Microcentrifuge Floating Racks, Bel-Art, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F18875-6411 F18875-6401
16126-544BG 57.29 USD InStock
16126-544 13197-152
SP Bel-Art Round Microcentrifuge Floating Racks, Bel-Art, Bel-Art Products, a part of SP
Racks Tube Racks
Molded of steam autoclavable polypropylene, these racks fit 400 mL or 1000 mL beakers.
  • Floating rack design
  • Easy to unload
  • Polypropylene construction
  • Autoclavable
Tubes remain completely immersed in a beaker while tops rest above the rack. Unload by lightly pressing down against the bench until the legs touch, releasing the tubes. Choose from 8- or 20-place racks with 10.8 mm holes for 1.5 mL microtubes. Racks are 6.4 mm (¹/₄") thick, with 19 mm (³/₄") long legs and a center handle.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty