Would you like to log in again?
SP Bel-Art Precisionware® Bottles, Wide Mouth, HDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F10625-0018 106250005 10625-9007 F10625-0006 10625-9005 10625-9018 F10625-0007 10625-9006
12000-121CS 345.29 USD InStock
12000-121 16126-759 47748-112 47745-338 47748-114 16126-760 47750-620 47748-110
SP Bel-Art Precisionware® Bottles, Wide Mouth, HDPE, Bel-Art Products, a part of SP
Bottles Plastic Bottles
Rigid high-density polyethylene, laboratory/pharmaceutical grade bottles are leakproof and supplied with matched polypropylene closures.
  • Rigid HDPE construction
  • Polypropylene closures
  • Leakproof design
  • Recycling code molded into the base
Constructed from FDA compliant material, the wide mouth design allows for easy filling and pouring. The design also simplifies the packaging and storing of liquids and powders. Bottles 250 ml and larger feature a recycling code molded into the base.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty