Would you like to log in again?
SP Bel-Art Gallon Bottle, Wide-Mouth, Bel-Art Products, a part of SP
Supplier: Bel-Art
F10638-1010
16126-764CS 99.28 USD InStock
16126-764
SP Bel-Art Gallon Bottle, Wide-Mouth, Bel-Art Products, a part of SP
Bottles Plastic Bottles
High-density polyethylene bottle features a foam-lined, white polypropylene cap and offers excellent chemical and impact resistance.
  • HDPE construction
  • Foam-lined polypropylene cap
  • Chemically resistant
Wide mouth facilitates pouring and refilling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty