Would you like to log in again?
SP Bel-Art Polystormer™ Square Edge Bottles, Wide Mouth, Bel-Art Products, a part of SP
Supplier: Bel-Art
F10640-0004 F10640-0032 F10640-0008 F10640-0016
30617-598CS 266.37 USD InStock
30617-598 16126-776 16126-780 16126-772
SP Bel-Art Polystormer™ Square Edge Bottles, Wide Mouth, Bel-Art Products, a part of SP
Bottles Plastic Bottles
These high-density polyethylene bottles with polypropylene screw closures store more samples or chemicals in limited shelf space than round bottles.
  • HDPE bottle construction
  • Polypropylene screw closures
  • Square shape stores more and is easier to ship
Their shape makes them ideal shipping containers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty