Would you like to log in again?
Nalgene® Laboratory Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
2106-0001 2106-0002 2104-0002 2106-0008 2106-0004 2104-0016 2104-0032 2104-0001 2104-0048 2104-0004 2106-0032 2104-0008 2106-0016
73520-408CS 335.04 USD InStock
73520-408 16127-226 16127-204 16125-107 16125-027 16125-005 16125-049 16127-124 16127-146 16127-168 16125-082 16125-060 16127-180
Nalgene® Laboratory Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Leakproof, translucent lab quality bottles with many applications in the field or laboratory.
  • Bottles are available in both translucent and amber finishes
  • Translucence allows easy viewing of liquid levels
  • Amber reduces UV light transmissions to protect light-sensitive liquids
  • Excellent durability and sample protection
Bottles are manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements. Bottles are supplied with polypropylene screw closures.
Suitable for anything from wide general lab use to storing critical, high-value materials. Bottles are more rigid than LDPE, with excellent chemical resistance to most acids, bases, and alcohols. The wide mouth is easy to fill with dry materials or liquids. Suitable for freezer use to –100 °C(–148 °F).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty