Would you like to log in again?
Nalgene® Laboratory Bottles, Teflon® FEP, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2100-0016 2100-0032 2100-0008 2100-0004
16128-535CS 1472.64 USD InStock
16128-535 16128-499 16128-455 16128-411
Nalgene® Laboratory Bottles, Teflon® FEP, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Laboratory bottles offer extraordinary chemical and temperature resistance.
  • Ideal for high or low temperature work, trace metal analysis, and applications with organic solvents
  • Bottle with cap withstands temperatures from –105 to 150 °C (–157 to 302 °F)
  • Clear Tefzel® ETFE closures
  • Leakproof
  • Autoclavable
Note: Before autoclaving, set closure on top of bottle without engaging threads.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty