Would you like to log in again?
Chemware® Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Supplier: Saint-Gobain
Chemware®
D1069589 D1069591 D1069592 D1069593 D1069590
16128-540EA 223.25 USD InStock
16128-540 16128-541 16128-539 16128-542 16128-543
Chemware® Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Bottles Plastic Bottles
Good for trace analysis. Leakproof, unbreakable PFA bottles for storage of virtually all corrosive chemicals including HF, nitric and perchloric acids.
  • Bottles withstand temperatures from 500 °F (260 °C) to the cryogenic range without becoming brittle
  • Chemically inert, non-stick surface is easy to clean
  • PFA cap
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty