Would you like to log in again?
SP Bel-Art Mason Jars, Wide Mouth, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F10916-0000 F10914-0000 F10913-0000 F10917-0000 F10915-0000
16128-660CS 371.17 USD InStock
16128-660 16128-626 16128-604 16128-648 70690-010
SP Bel-Art Mason Jars, Wide Mouth, Polypropylene, Bel-Art Products, a part of SP
Jars General Use Jars
Highly shock-resistant polypropylene, heavy-walled Mason Jars are ideal for industrial waste sampling, long-term storage, and a variety of laboratory uses.
  • Polypropylene construction
  • Shock-resistant
  • Heavy-walled design
  • Autoclavable
They have standard 70 mm, G-threaded necks, and screw caps with unique grip lugs and a removable leakproof silicone rubber ring seal.
Cap and jar are steam autoclavable at 121°C (250°F). Loosen cap when autoclaving.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty