Would you like to log in again?
Nalgene® Laboratory Bottles, Polypropylene Copolymer, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2105-0016 2105-0001 2105-0004 2105-0008 2105-0032 2105-0002
16129-061CS 366.18 USD InStock
16129-061 16129-040 16129-006 16129-028 16128-977 16128-954
Nalgene® Laboratory Bottles, Polypropylene Copolymer, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Leakproof lab quality bottles for chemicals, specimens, and materials where autoclaving is required.
  • Excellent durability and sample protection
  • Excellent chemical resistance to most acids, bases, and alcohols
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Translucent bottles offer better clarity than LDPE or HDPE. The wide mouth is easy to fill with dry materials or liquids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty