Would you like to log in again?
Nalgene® PMP Jars, Straight-Sided, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2117-1000 2117-0060 2117-0125 2117-0250 2117-0500
16129-390CS 346.56 USD InStock
16129-390 16129-414 16129-354 16129-356 16129-378
Nalgene® PMP Jars, Straight-Sided, Thermo Scientific
Jars General Use Jars
Transparent, autoclavable, with straight sides
Top is recessed to allow stacking. Jars of 125mL and 250mL sizes have standard mason jar threads. Ideal for museum jars — formaldehyde loss only about 2% per year. Remove closure before autoclaving, or set closure on top of jar at an angle without engaging the threads.
Suitable for food and beverage use. Meets requirements of CFR21, Part 177.1520 section of Food Additives Amendment of Federal Food and Drug Act.
Polypropylene white screw closures are linerless, noncontaminating, and autoclavable.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty