Would you like to log in again?
Nalgene® Bottles and Carboys, Fluorinated High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
2097-0016 2097-0020 2097-0050 2097-0032 2097-0008 2097-0005 2097-0010
16129-708CS 339.53 USD InStock
16129-708 16129-704 16129-706 16129-710 16129-700 16129-712 16129-702
Nalgene® Bottles and Carboys, Fluorinated High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Safe, durable, cost-efficient line of narrow mouth bottles and carboys feature fluorinated surfaces (inside and outside), for improved barrier properties and reduced solvent absorption and penetration.
  • Split- and puncture-resistant
  • With polypropylene screw closure
  • Heavy-duty walls, Leakproof
  • 10 and 20 L sizes feature molded-in graduations
Fluorination enhances long-term container performance and prevents or reduces permeation loss. Useful with most aggressive organic solvents.
Ordering information: Closures not available separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty