Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREXPLUS® Safety-Coated Solution Bottles, Corning

Supplier: Corning
PYREXPLUS®
61596-19L 61596-13L
16145-166CS 983.85 USD InStock
16145-166 16145-164
PYREXPLUS® Safety-Coated Solution Bottles, Corning
Bottles Glass Bottles
PYREX® borosilicate glass, coated.

  • Conventional bottle shape
  • Autoclavable at 121 °C and resistant to thermal shock
  • Used for storage of solutions, ideal for media preparation

Designed with protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Neck is tooled to increase mechanical strength and for uniform stopper fit.

Caution: Do not place over direct heat or flame.

Graduated at 500 ml increments and fits rubber stopper No. 12.

Can be written on with any standard lab marker, permanent or water-soluble

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty