Would you like to log in again?
WHEATON® Media Square™ Media Bottles, PET, Graduated, DWK Life Sciences
Supplier: DWK Life Sciences
Wheaton Media Square™
219975
16149-167CS 285.26 USD InStock
16149-167
WHEATON® Media Square™ Media Bottles, PET, Graduated, DWK Life Sciences
Bottles Cell Culture Bottles
Sterile, lightweight bottles offer the clarity of glass with the strength of polyethylene terephthalate.
  • No-drip pour lip offers clean, fast, easy pouring
  • Polypropylene caps are lined with foamed polyethylene
Tested superior for pH stability, temperature durability, and cloning efficiency. Permanent molded-in graduations provide accurate volume determination at a glance.
Ordering information: Bottles with caps attached are packed in convenient shrink-wrapped trays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty