Would you like to log in again?
Safety-Coated Reagent Bottles, Amber, Narrow Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
220955 220935 220956 220976 220945 220924 220926 220925 220936 220975 220946
16150-308CS 292.26 USD InStock
16150-308 16150-393 16150-402 16150-070 16150-068 16150-058 16150-048 16150-400 16150-394 16150-341 16150-056
Safety-Coated Reagent Bottles, Amber, Narrow Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Create a safe work environment with Wheaton safety coated bottles.
Bottles with caps are used for distilled water, analytical standards and reagents. Metal foil liners are generally more resistant to organic solvents.
The strength, slip resistance and impact absorption qualities of the plastisol safety coating reduce potential breakage and exposure to hazardous materials. Wheaton '910' soda-lime flint amber glass protects light-sensitive samples.
If a bottle is accidentally broken, the PVC based plastisol coating on the outside of bottle retains the contents long enough for proper disposal. Ideal for lab or field applications, the bottle’s non-slip coating allows for greater ease and stability when handling wet or dry. The narrow Boston Round style is ideal when used for liquid. The bottles are manufactured from soda lime glass that conforms to USP Type III requirements.
Ordering information: Available with caps attached or bulk packs without caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty