Would you like to log in again?
Safety Coated Reagent Bottle, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
221017 221014
16150-082CS 158.57 USD InStock
16150-082 16150-080
Safety Coated Reagent Bottle, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Safety coated narrow neck reagent bottle for the safe storage of laboratory samples.
  • Plastisol coated exterior of bottle retains contents if bottle is accidentally broken
  • Made from low extractable clear borosilicate glass
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • Supplied with attached white polypropylene 33-430 cap with PTFE liner
  • Autoclavable
Certifications: It conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty