Would you like to log in again?
Nasal Spray Bottles, Qorpak
Supplier: Qorpak
PLC-03662
16151-814CS 78.23 USD InStock
16151-814
Nasal Spray Bottles, Qorpak
Bottles Dropping and Dispensing Bottles
Nasal spray mist bottles are made of squeezable, resilient LDPE and are supplied with the spray plug and overcap.
They can also be used as dropper bottles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty