Would you like to log in again?
Safety-Coated Boston Round Bottles, Amber, Qorpak
Supplier: Qorpak
GLC-07106 GLC-04920 GLC-08947 GLC-07019 GLC-02268 GLC-07105 GLA-00955 GLA-00952 GLA-00953 GLC-02278 GLC-02270 GLC-02271 GLC-02276 GLC-02261 GLC-02256 GLA-00959 GLC-02272 GLA-00951 GLC-02251 GLC-02249 GLA-00954 GLC-06989 GLC-02262 GLC-02255 GLC-02250 GLA-08639 GLC-07407 GLC-02253 GLC-02279 GLC-07018 GLC-02274 GLC-02265 GLC-02258 GLA-00958 GLA-00956 GLC-02260
16150-912CS 319.77 USD InStock
16150-912 16152-302 16150-936 16150-915 16150-937 16152-304 16150-930 16150-931 16152-306 16150-954 93000-932 93000-954 16150-933 16150-918 11311-378 89204-842 11311-308 16150-950 16150-951 16152-300 93000-926 16150-901 16150-924 16150-904 16150-921 16150-900 16151-594 11311-542 16150-927 16150-905 89184-908 89234-734 89234-736 89021-872 93001-646 93000-930
Safety-Coated Boston Round Bottles, Amber, Qorpak
Bottles Glass Bottles
Safety-coated amber Boston round bottles protect samples from UV rays and are ideal for light sensitive products.
  • Safety-coated boston round bottles are made of glass
If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal. These bottles are slip- and impact-resistant, making them the perfect bottle for handling volatile substances.
Ordering information: Bottles 89204-842, 93000-926, 93000-930, 93000-932, and 93000-954 are supplied Vacuum treated and Ionized. Bottle 89021-872 is supplied pre-cleaned for use with volatiles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty