Would you like to log in again?
Polypropylene Screw Caps, White, Wheaton
Supplier: DWK Life Sciences
239273 239274 242224 239281 W238520-2400 W238520-1525 W238520-2000 239290 239287 239218 242226 239203 W238515-1800 242220 239283 239226 239227 239286 242218 W238520-1800 239209 239229 239225 242216 239284 W238515-2400 239239 242217 239289 242212 239288 239235 W238520-2200 242225 239285 239217 239292 239201 239275 242210 W238520-1325 242228 239231 239233 242222
10796-432CS 58.03 USD InStock
10796-432 16299-294 16299-296 16299-290 10796-436 10796-434 16153-227 16153-203 16153-225 16153-289 16153-201 16153-224 16153-287 16153-288 16153-285 16153-286 16153-283 16153-284 16153-281 14221-572 14221-518 14221-574 16299-298 16299-310 16153-218 30620-984 30620-982 16153-239 14221-522 14221-602 16153-274 16153-275 16153-273 16299-306 16153-292 16299-308 16153-290 80082-950 14221-508 16299-302 16299-304 14221-506 14221-588 16299-288 16299-300
Polypropylene Screw Caps, White, Wheaton
Closures Bottle Closures
These caps are designed for use with WHEATON® glass bottles or vials.
  • White surface for writing on cap
  • Offered in a variety of liner materials
Polyethylene foam provides compressibility and sealing properties, and is excellent for use with environmental samples, pharmaceuticals, and diagnostic reagents. The Poly-vinyl liner provides an excellent moisture barrier, and is good for use with mild acids, alkalis, solvents, alcohols, oils, and aqueous products. The PTFE-faced foamed PE liner is ideal for use with high purity chemicals, strong acids, and solvents.
White polypropylene screw caps feature a clear surface for writing on cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty