Would you like to log in again?
WHEATON® Polypropylene Screw Cap, Linerless, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
239506-02 239510-03 239501-02 239501 239501-03 239510-02 239510-04 239512-02 239516-02 239506-06 239501-06 239512-04 239510 239512 239506 239506-04 239506-03 239516-03 239516-04 239510-06 239516
16153-509CS 124.22 USD InStock
16153-509 16153-507 16153-505 16153-503 16153-467 16153-489 16153-479 16153-487 16153-465 16153-499 16153-477 16153-463 16153-485 16153-475 16153-483 16153-495 16153-473 16153-481 16153-471 16153-493 16153-491
WHEATON® Polypropylene Screw Cap, Linerless, DWK Life Sciences
Closures Bottle Closures
A variety of polypropylene caps are offered separately for Wheaton Leak Resistant Narrow Mouth Bottles
Caps can be autoclaved for 15 minutes at 121ºC at 15 psi. Polypropylene caps are available in natural, white, red, blue and green
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty