Would you like to log in again?
Wide Mouth Blake Packer, HDPE, Natural, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
209685 209683 209686
16153-527CS 382.82 USD InStock
16153-527 16153-533 16153-531
Wide Mouth Blake Packer, HDPE, Natural, WHEATON®, DWK Life Sciences
Bottles Plastic Bottles
Wheaton wide mouth blake packers are manufactured from high density polyethylene.
High Density Polyethylene (HDPE) Characteristics: Flexible but more rigid than LDPE. Natural color is milky white, semi-translucent depending on density. White HDPE helps protect light sensitive samples. Good impact strength and stress crack resistance. Good chemical resistance. Good vapor barrier but poor gas barrier. Can be sterilized using ethylene oxide (EtO) or gamma radiation.
The wide mouth is ideal for solid samples. Pre-attached white polypropylene screw cap with foamed polyethylene liner provides cleanliness. Blake design maximizes storage space.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty