Would you like to log in again?
Media Lab Bottle, Non-Graduated, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
219497 W219500 219575 219885 219495 219417 W219580 W219580 219577 W219420 219579 219415 219419 219499
14216-222CS 351.69 USD InStock
14216-222 89200-074 89200-076 66012-903 16158-081 14231-278 16154-718 16154-715 14231-290 14231-280 89200-070 16364-984 16152-154 16364-982
Media Lab Bottle, Non-Graduated, WHEATON®, DWK Life Sciences
Bottles Cell Culture Bottles
Wheaton media bottles are manufactured from Wheaton 400 borosilicate glass that conforms to USP Type I requirements and have a low alkali content to help prevent changes in pH to maintain the purity of its contents.
  • Hand grips on the 1 L bottle makes for easy handling
The pour lip and sloped shoulders facilitate pouring and cleaning. Pre-attached black phenolic screw caps with liner are attached to each bottle.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty