Would you like to log in again?
Leak-Resistant Bottles, Amber, High-Density Polyethylene, Narrow Mouth, WHEATON®
Supplier: DWK Life Sciences
209127 209128 209129 209124 209125 209123 209130 209121 209126
16155-121CS 175.33 USD InStock
16155-121 16155-130 16155-124 16155-125 16155-123 16155-128 16155-129 16155-126 16155-127
Leak-Resistant Bottles, Amber, High-Density Polyethylene, Narrow Mouth, WHEATON®
Bottles Plastic Bottles
Ideal storage bottles.
  • Amber in color to protect light-sensitive samples
  • Double-seal closure for leak-resistance
Designed with a special pour lip to facilitate safe, clean dispensing.
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty