Would you like to log in again?
Leak-Resistant Bottles, Low-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences
209427 209425 209428
16155-428CS 460.62 USD InStock
16155-428 16155-427 16155-425
Leak-Resistant Bottles, Low-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Bottles Plastic Bottles
Lightweight bottles are ideal for general laboratory use.
  • Wide mouth facilitates filling and pouring
  • Bottles are designed with a special pour lip to facilitate safe, clean dispensing
  • Double-seal closure for leak resistance
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty