Would you like to log in again?
Media Lab Bottles, Amber, Graduated, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
219845 219855 219865
16149-170CS 1083.24 USD InStock
16149-170 16159-670 16159-674
Media Lab Bottles, Amber, Graduated, WHEATON®, DWK Life Sciences
Bottles Cell Culture Bottles
Wheaton media bottles are manufactured from Wheaton 400 borosilicate glass.
  • Media lab bottles have 25 ml graduations and a writing patch
The glass conforms to USP Type I requirements and has a low alkali content to help prevent changes in pH. This also helps to maintain the purity of its contents. The pour lip and sloped shoulders facilitate pouring and cleaning. Pre-attached black phenolic screw caps with liner are attached to each bottle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty