Would you like to log in again?
WHEATON® Black Plastic Caps for Media Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
240281 240421 240223 W013743 240321 240264 W013735 W240406 240415 240463 W013736 W013737 240280 240416 240381 240228 240080 240419 240081 W013741 240481 240330 240408 240414 W013739 240269 240409 240480 240319
16160-120CS 583.36 USD InStock
16160-120 16299-230 16160-323 16160-301 16160-720 80090-054 80090-056 80090-058 80090-052 74830-130 66012-353 66022-800 16160-505 16160-527 16160-108 66012-394 66012-350 16299-234 66012-372 16299-240 80090-064 66012-419 80090-048 25457-039 14007-174 16299-228 16299-224 16299-246 16299-248
WHEATON® Black Plastic Caps for Media Bottles, DWK Life Sciences
Closures Bottle Closures
Solid-top lined caps are suitable for a variety of applications
Choose from rubber-, fluoropolymer resin-, polyethylene-, or metal foil-lined caps. Rubber-lined caps are for culture work. Fluoropolymer resin-lined caps are for maximum product safety. Both rubber- and fluoropolymer resin-lined caps are autoclavable. Polyethylene-lined caps are designed for distilled water, analytical standards, and reagents. Metal foil-lined caps are for organic solvents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty