Would you like to log in again?
Serum Bottle, Borosilicate Glass, Aluminum Seal, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences
61000G-200 61000G-20 61000G-30 61000G-100 61000G-50 61000G-60 61000G-10 61000G-5 61000G-125
16161-937CS 471.75 USD InStock
16161-937 16161-936 16161-935 16161-934 16161-933 16161-932 16161-931 89004-554 16161-938
Serum Bottle, Borosilicate Glass, Aluminum Seal, KIMBLE®, DWK Life Sciences
Bottles Glass Bottles
Kimble serum bottle is constructed from a unique borosilicate glass that offers the highest known resistance to chemical attack.
  • GPI neck finish: 20A
Ideal for packaging, storage, and application of injectable and parenteral solution requiring the utmost purity. Bottle also offers a high resistance to thermal and mechanical shock.
Certifications: Meets or exceeds USP Specifications for Type I Glass Containers.
Ordering information: Bottle is not supplied with a closure.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty