Would you like to log in again?
KIMBLE® KIMAX® Media/Storage Bottles, KG-35 Borosilicate Glass, Graduated, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
61111T-1000 61110R-500 61110T-250 61110R-1000 61110P-250 61110P-125 61110R-125 61110T-1000 61110P-500 61110P-1000 61110R-250 61110T-125 61110T-500 61111R-1000 61111P-1000
16171-014CS 260.2 USD InStock
16171-014 16171-004 16171-016 16171-006 16171-010 16171-000 72630-010 16171-002 66014-002 66014-004 66014-006 66014-008 66014-010 16171-008 66014-000
KIMBLE® KIMAX® Media/Storage Bottles, KG-35 Borosilicate Glass, Graduated, DWK Life Sciences
Bottles Cell Culture Bottles
Screw thread graduated bottles come assembled with autoclavable closures.
  • All bottles feature a large white marking spot
Choose from closures that are black phenolic 14-B white rubber-lined, black phenolic PTFE-faced 14-B white rubber-lined, or black polypropylene welded to a PTFE/silicone liner. The PTFE/silicone liner eliminates the possibility of glue contamination to bottle contents.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty