Would you like to log in again?
Serum Bottles, Borosilicate Glass, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
223739 223762 223760 223761 223743 223763 223764 223742 223745 223747 223746 223738 223748 223766 W223712
16171-466CS 633.75 USD InStock
16171-466 16171-433 16171-499 16171-401 89422-806 16171-330 16171-341 16171-352 16171-374 16171-385 16171-320 16171-319 16171-308 16171-524 16171-557
Serum Bottles, Borosilicate Glass, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Serum bottles are ideal for long and short term sample storage, lyophilization, and as vaccine/injectable drug containers.
  • Bottles fit most lyophilization applications
Clear bottles are manufactured from Wheaton 400 borosilicate molded glass, and conform to USP Type I requirements. Amber bottles are manufactured from Wheaton 500 borosilicate glass, and conform to USP Type I requirements. All bottles feature a 13Int.Ø×20Ext.Ø mm mouth opening, with the exception of 89422-806 which is 7Int.Ø×13Ext.Ø mm.
Ordering information: Bottles are packaged in shrink-wrapped, partitioned modules to reduce breakage.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty