Would you like to log in again?
Serum Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
223952 223950
16171-567CS 275.32 USD InStock
16171-567 16171-420
Serum Bottles, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Serum graduated bottles accepts standard stoppers and seals.
  • Manufactured from 400 borosilicate glass that conforms to USP Type I requirements
  • Mouth Int.Ø×Ext.Ø: 15×30 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty