Would you like to log in again?
Nalgene® Economy Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2089-0032 2089-0016 DS2085-0016 2089-0001 2089-0002 DS2085-0004 2089-0008 2089-0004 DS2085-0032
16172-110CS 205.44 USD InStock
16172-110 16172-122 16172-130 16172-120 16172-104 16172-126 16172-124 16172-108 16172-128
Nalgene® Economy Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Economical, single-use, break-resistant bottles feature thinner walls and are more flexible than original Nalgene® bottles.
  • Excellent chemical resistance to most acids, bases, and alcohols
  • Guaranteed leakproof
  • Available in natural or amber
  • Narrow mouths are ideal for pouring
Ideal for short-term, general-purpose laboratory and field applications, such as collecting samples, or for storing working solutions like buffers, bench reagents, and diluted chemical aliquots. Natural bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Certifications: Amber bottles comply with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Delivery information: Supplied with polypropylene screw closures.
Caution: Not recommended for hazardous and critical material containment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty