Would you like to log in again?
Nalgene® Economy Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
2189-0032 DS2185-0008 2189-0016 DS2185-0016 2189-0008 2189-0001 DS2185-0032 2189-0004 DS2185-0004 2189-0002
16172-166CS 178.56 USD InStock
16172-166 16172-144 16172-164 16172-148 16172-146 16172-168 16172-150 16172-162 16172-170 16172-160
Nalgene® Economy Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Economical, single-use, break-resistant bottles feature thinner walls and are more flexible than original Nalgene® bottles.
  • Excellent chemical resistance to most acids, bases, and alcohols
  • Available in natural and amber finishes
  • Guaranteed leakproof
Ideal for short-term, general-purpose laboratory and field applications, such as collecting samples, or for storing working solutions like buffers, bench reagents, and diluted chemical aliquots. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Certifications: Amber bottles comply with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Delivery information: Supplied with polypropylene screw closures.
Caution: Not recommended for hazardous and critical material containment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty