Would you like to log in again?
Nalgene® Wide Mouth Rectangular Bottles, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2007-0032 2007-0004 2007-0016 2009-0008 2009-0016 2007-0064 2009-0004 2009-0064 2009-0032 2007-0008
16186-291CS 427.2 USD InStock
16186-291 16186-338 16186-316 16186-258 16186-112 16186-134 16186-054 16186-098 16186-076 16186-270
Nalgene® Wide Mouth Rectangular Bottles, High-Density Polyethylene, Thermo Scientific
Bottles Plastic Bottles
These rectangular bottles take up less shelf space than Boston round bottles.
  • The sturdy wall construction, straight sides, and wide mouth allow easy filling
  • Ideal for sampling, shipping, or storing liquids or dry materials
  • Excellent chemical resistance to most acids, bases, and alcohols
  • Suitable for freezer storage to –100 °C (–148 °F)
Bottles are available in both translucent and amber finishes. Translucence allows easy viewing of liquid levels. Amber reduces UV light transmissions to protect light-sensitive liquids.
Delivery information: Polypropylene screw closures are included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty