These flint glass bottles are excellent for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents.
- Graduated in ml and ounces
- Mix, measure, and store in the same container
They are designed with a slight shoulder for easy pouring and handling.
Ordering information: Bottles 36319-325, 36319-327, 36319-329, 36319-330, and 36319-319 are supplied pre-cleaned for volatiles.