Would you like to log in again?
Straight Sided Rounds, Wide Mouth, Qorpak®
Supplier: Qorpak
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GLA-00867 GLC-05575 GLC-01665 GLC-01584 GLC-01700 GLC-01677 272171 GLC-01623 239229 GLC-01693 GLC-04910 GLC-01681 239228 GLC-01595 GLC-01684 GLC-01717 GLA-00850 GLA-00864 GLC-01629 GLC-01675 239225 GLC-01692 GLC-01685 GLC-01596 GLC-01673 GLC-07897 GLC-01628 GLA-00853 GLC-01586 GLA-00861 275736 GLC-01716 239227 GLC-01664 GLC-01687 GLC-01634 GLC-01605 250240 272173 GLC-01728 GLC-01672 GLC-01731 GLC-01582 GLC-01612 GLC-01705 GLC-01635 GLC-01624 GLA-00857 GLC-01606 239230 GLC-01721 GLC-07896 GLC-01627 GLC-01715 GLA-00854 GLC-01725 272174 GLC-01581 GLC-01666 GLC-01642 GLC-01592 GLC-08640 GLC-01620 GLC-01724 GLC-01676 GLC-07451 250239 239224 GLC-01732 272170 272172 GLA-00859 GLC-01703 272175 GLC-01720 239223 GLC-01679 GLC-01611 GLC-01686 GLC-04875
16195-102CS 178.17 USD InStock
16195-102 16195-340 16195-022 16195-063 16195-383 16195-061 16151-580 16054-142 16194-100 16054-140 16054-182 16054-148 16054-146 76170-396 16195-306 16195-747 16195-703 16195-667 10847-280 76170-406 16195-059 76170-408 16195-771 16195-772 16195-055 16195-770 36319-145 82027-766 76419-796 36319-103 36319-147 76170-400 16054-176 76419-798 76170-402 76170-404 76419-794 16218-178 76419-804 16195-088 11311-390 16195-044 76419-800 76419-802 16195-361 82024-648 36319-232 36319-111 76170-410 36319-113 36319-151 36319-231 16218-176 89204-942 36319-149 16195-408 16195-727 16081-040 16195-328 16195-725 16195-769 16195-689 11311-142 11311-386 16195-033 16218-180 16218-182 76170-422 14002-387 11311-388 11311-662 11311-148 36319-237 16194-518 16087-308 76170-386 10847-230 76487-202 76487-206 76487-204
Straight Sided Rounds, Wide Mouth, Qorpak®
Jars General Use Jars
Clear, flint glass straight sided round jars are designed without shoulders for maximum storage capacity and content visibility.
  • Made from Type III Soda Lime Glass
  • USP certified Pharma grade glass
  • Clear glass is appropriate when visibility is important and protection from UV rays is not necessary
The wide mouth opening is ideal for mixing, storing, and sampling. Many closures are available to suit particular needs.
Ordering information: Bottles 36319-113, 36319-145, 36319-147, 36319-149, 36319-151, and 16151-580 are supplied pre-cleaned for volatiles. Bottle 36319-103 is supplied pre-cleaned and certified for volatiles. Bottle 82024-648 is supplied pre-cleaned for semi-volatiles. Bottles 16054-176, 16054-182, and 14002-387 are supplied pre-cleaned and certified for semi-volatiles.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty