Would you like to log in again?
Safety Coated Jar, Clear, Wide Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
216629 216637 216639 216627
16197-126CS 124.81 USD InStock
16197-126 16197-116 16197-128 16197-118
Safety Coated Jar, Clear, Wide Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Create a safe work environment with safety coated jars.
If the jar is accidentally broken, the PVC based plastisol coating on the outside of jar retains the contents long enough for proper disposal. Ideal for lab or field applications, the jar's non-slip coating allows for greater ease and stability when handling wet or dry. The wide mouth is ideal for liquid. The jars are manufactured from clear, soda lime glass that conforms to USP Type III requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty