Would you like to log in again?
KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
Supplier: DWK Life Sciences
KIM-KAP®
73660-25 73665-20 73664-20 73664-13 73662-13 73664-25 73660-38 73662-25 73662-20 73663-20 73660-18 73662-18 73664-16 73663-13 73665-25 73665-16 73660-20 73662-16 73665-18 73660-16 73663-16 73663-25 73660-13
60825-815CS 226.64 USD InStock
60825-815 16199-021 16199-010 16199-011 16199-022 16199-020 16199-018 16199-007 16199-019 16199-008 16199-005 16199-016 16199-017 16199-003 16199-014 16199-015 16199-004 16199-002 60825-801 60825-812 60825-798 60825-787 16199-009
KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
Closures Test/Sample Tube Closures
These KIM caps are designed to fit culture tubes with outer diameters ranging from 13 mm to 38 mm.
  • Autoclavable
  • Color coded
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty