Would you like to log in again?
Dropping Bottles, Amber, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences
15040P-60 15040G-60 15040P-30 15040G-30
16199-039CS 60.22 USD InStock
16199-039 16199-037 16199-034 16199-031
Dropping Bottles, Amber, Kimble®, DWK Life Sciences
Bottles Dropping and Dispensing Bottles
ASTM E438, Type III amber glass protects light-sensitive chemicals.
Supplied with 20-400 GPI screw cap, bulb and dropper assembled on the bottle. Dropper is available in polyethylene or USP Type III glass.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty