Would you like to log in again?
Black Phenolic Screw Caps, Pulp/Tinfoil Liner, Qorpak®
Supplier: Qorpak
CAP-00215 CAP-00314 CAP-00292
16199-668CS 1096.06 USD InStock
16199-668 16199-840 16199-805
Black Phenolic Screw Caps, Pulp/Tinfoil Liner, Qorpak®
Closures Bottle Closures
Pulp/Tin Foil lined caps offer good chemical resistance and are typically used with solids and powders
These black phenolic screw caps are made of phenolic resin.
Good resistance to hydro-carbons, alcohols, ketones and oils. Not
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty