Would you like to log in again?
Biohazard Wipes and Liners, Current Technologies
Supplier: Current Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BH92910 BH90044 BH44985 BH91033 BH 16016 ET BH32000 BH44200 BH33400 BH11016ET BH91616 BH 10600 BH16100 BH93016 BH91625
16200-212CS 345.19 USD InStock
16200-212 16200-210 82030-864 16200-204 16200-216 10147-842 16200-214 10147-840 16200-208 16200-206 89092-532 89092-534 10147-844 10147-848
Biohazard Wipes and Liners, Current Technologies
Wipes Disposable Wipes and Towels Disinfectant Wipes
Low-linting wipes and liners are specifically designed to protect workers and labs from contamination from blood, body fluids, tissue, specimens plus chemicals.
Wipes and liners are available in several sizes and in large rolls for cutting to size. Wipes are also available in standard absorbency, which can absorb 9× its own weight, heavy absorbency, which can absorb 12× its weight and super absorbency, which can absorb 25× its weight.
Specifically sized dispensers are constructed of clear acrylic.
Absorbent white side soaks up blood, tissue, specimen, body fluids, and chemicals. Bright orange barrier side, made of coextruded plastic, blocks soak-through to gloves, hands and counters, limiting the spread of contamination. Wipes are impermeable to strong solvents including xylene and xylene substitutes, acetone, ketones, alcohols, and formalin. Use to pop caps, wipe pipets and probes, handle specimens and line benches, counters and trays, clean up spills.
Ordering information: Bio-Screen wipes and liners are non-sterile and are not for use on patients.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty