Would you like to log in again?
Nalgene® Unitary™ Vented Safety Wash Bottles, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene® Unitary™
2436-1001 2436-1005 2436-0501 2436-0502 2436-0504 2436-0503 2436-0506 2436-0505
16210-921CS 364.04 USD InStock
16210-921 16210-919 16210-917 16210-927 16210-915 16210-925 16210-935 16210-923
Nalgene® Unitary™ Vented Safety Wash Bottles, Low-Density Polyethylene, Thermo Scientific
Bottles Wash Bottles
LDPE bottles feature a unique vent system to keep hazardous volatile liquids in the bottle and off the lab bench.
Sodium hypochlorite (bleach) bottles are supplied with vented, white HDPE 38 mm closure; all other bottles supplied with vented polypropylene 38 mm colored closure. To prevent cross-contamination, each bottle has a color bar below the chemical name that corresponds to the colored closure.
Unitary design prevents contamination when filling the bottle. A strong, steady stream of liquid is dispensed when the bottle is squeezed.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty