Would you like to log in again?
Nalgene® Heavy-Duty Vacuum Bottles, Polypropylene Copolymer, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2126-2000 2126-4000 2126-5000 DS2126-0250 2126-1000
16056-970CS 167.04 USD InStock
16056-970 16331-100 16210-078 16210-076 16211-080
Nalgene® Heavy-Duty Vacuum Bottles, Polypropylene Copolymer, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Sturdy, thick-walled bottles are designed for heavy-duty applications in the biotechnology lab.
  • Rugged caps with TPE gaskets guarantee leakproof service under vacuum
  • Autoclavable
Bottles withstand repeated application of full vacuum for 24-hour periods at room temperature (20 °C, 68 °F). Use as waste aspirator bottle (with Barbed Bulkhead Fittings 16225-232) or as a vessel for scale-up activities in cell culture applications.
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: For replacement caps, see 16219-122 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty